Assay Details
Gene Symbol: Capns1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017118.1 | NP_058814.1 | 4 - 5 | 389 | 97 | |
GenBank mRNA | BC098068.1 | - | 4 - 5 | 389 | 97 | |
FQ229527.1 | - | 72 | 97 | |||
U10861.1 | - | 1 - 2 | 78 | 97 | ||
U53859.1 | - | 3 - 4 | 325 | 97 |
Gene Symbol: LOC100911363 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003748836.3 | XP_003748884.1 | 4 - 5 | 438 | 97 | |
XM_008759263.1 | XP_008757485.1 | 4 - 5 | 640 | 97 |
Gene Symbol: LOC100912380 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006228796.2 | XP_006228858.2 | 3 - 4 | 165 | 97 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
