Assay Details
Gene Symbol: Nr1h2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031626.1 | NP_113814.1 | 1 - 2 | 110 | 105 | |
GenBank mRNA | BC157812.1 | - | 1 - 2 | 89 | 105 | |
U14533.1 | - | 1 - 2 | 110 | 105 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC157812.1, NM_031626.1, U14533.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
