Assay Details
Gene Symbol: Runx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008768599.1 | XP_008766821.1 | 3 - 4 | 2735 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
