Assay Details
Gene Symbol: Cct8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105897.2 | NP_001099367.2 | 10 - 11 | 1224 | 81 | |
GenBank mRNA | CO400743.1 | - | 4 - 5 | 417 | 81 | |
FQ219784.1 | - | 2 - 3 | 111 | 81 | ||
FQ226011.1 | - | 10 - 11 | 1197 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
