Assay Details
Gene Symbol: Itgb1bp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106719.2 | NP_001100189.1 | 5 - 6 | 561 | 102 | |
XM_006239979.2 | XP_006240041.1 | 4 - 5 | 477 | 102 | ||
XM_006239980.2 | XP_006240042.1 | 5 - 6 | 506 | 102 | ||
XM_006239981.2 | XP_006240043.1 | 4 - 5 | 688 | 102 | ||
GenBank mRNA | BC169104.1 | - | 5 - 6 | 561 | 102 | |
FQ215419.1 | - | 4 - 5 | 396 | 102 | ||
FQ218542.1 | - | 4 - 5 | 438 | 102 | ||
FQ229091.1 | - | 5 - 6 | 564 | 102 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
