Assay Details
Gene Symbol: Itgb1bp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106719.2 | NP_001100189.1 | 7 - 8 | 802 | 74 | |
XM_006239979.2 | XP_006240041.1 | 6 - 7 | 718 | 74 | ||
XM_006239980.2 | XP_006240042.1 | 7 - 8 | 747 | 74 | ||
XM_006239981.2 | XP_006240043.1 | 6 - 7 | 929 | 74 | ||
GenBank mRNA | BC169104.1 | - | 7 - 8 | 802 | 74 | |
FQ229091.1 | - | 6 - 7 | 1306 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
