Assay Details
Gene Symbol: Hadhb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006239786.1 | XP_006239848.1 | 7 - 8 | 916 | 69 | |
XM_006239788.1 | XP_006239850.1 | 7 - 8 | 572 | 69 | ||
XM_006239789.2 | XP_006239851.1 | 7 - 8 | 847 | 69 | ||
GenBank mRNA | BC060545.1 | - | 7 - 8 | 499 | 69 | |
D16479.1 | - | 6 - 7 | 463 | 69 | ||
FM050540.1 | - | 7 - 8 | 527 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
