Assay Details
Gene Symbol: Hadhb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC060545.1 | - | 2 - 3 | 129 | 138 | |
FM050540.1 | - | 2 - 3 | 157 | 138 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC060545.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
