Assay Details
Gene Symbol: Ung | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013124.1 | NP_001013142.1 | 3 - 4 | 559 | 99 | |
XM_008769336.1 | XP_008767558.1 | 4 - 5 | 1230 | 99 | ||
GenBank mRNA | BC091211.1 | - | 3 - 4 | 559 | 99 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
