Assay Details
Gene Symbol: Wbscr22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135743.1 | NP_001129215.1 | 7 - 8 | 537 | 54 | |
GenBank mRNA | BC160891.1 | - | 7 - 8 | 537 | 54 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
