Assay Details
Gene Symbol: Fkbp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105922.1 | NP_001099392.1 | 1 - 2 | 95 | 73 | |
XM_008769130.1 | XP_008767352.1 | 1 - 2 | 114 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
