Assay Details
Gene Symbol: Rabgef1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108333.1 | NP_001101803.1 | 1 - 2 | 413 | 121 | |
XM_006249238.2 | XP_006249300.1 | 2 - 3 | 505 | 121 | ||
XM_006249239.2 | XP_006249301.1 | 2 - 3 | 257 | 121 | ||
XM_006249240.2 | XP_006249302.1 | 2 - 3 | 494 | 121 | ||
XM_008769150.1 | XP_008767372.1 | 2 - 3 | 504 | 121 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
