Assay Details
Gene Symbol: Sds | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053962.3 | NP_446414.3 | 1 - 2 | 53 | 67 | |
GenBank mRNA | BC088110.1 | - | 1 - 2 | 53 | 67 | |
Y00752.1 | - | 1 - 2 | 70 | 67 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC088110.1, NM_053962.3, Y00752.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
