Assay Details
Gene Symbol: Sds | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053962.3 | NP_446414.3 | 3 - 4 | 288 | 130 | |
XM_008769213.1 | XP_008767435.1 | 2 - 3 | 786 | 130 | ||
GenBank mRNA | BC088110.1 | - | 3 - 4 | 288 | 130 | |
J03863.1 | - | 3 - 4 | 378 | 238 | ||
Y00752.1 | - | 3 - 4 | 305 | 130 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
