Assay Details
Gene Symbol: Eif4h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006957.2 | NP_001006958.1 | 1 - 2 | 101 | 90 | |
XM_006249134.1 | XP_006249196.1 | 1 - 2 | 324 | 90 | ||
GenBank mRNA | BC083818.1 | - | 1 - 2 | 93 | 90 | |
CB718613.1 | - | 1 - 2 | 101 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
