Assay Details
Gene Symbol: Crcp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053670.3 | NP_446122.1 | 3 - 4 | 150 | 104 | |
GenBank mRNA | AF103950.1 | - | 1 - 2 | 65 | 104 | |
AF440799.2 | - | 3 - 4 | 150 | 104 | ||
BC059117.1 | - | 3 - 4 | 254 | 104 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
