Assay Details
Gene Symbol: Glt1d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008769205.1 | XP_008767427.1 | 3 - 4 | 593 | 119 | |
XM_222147.8 | XP_222147.4 | 3 - 4 | 448 | 119 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
