Assay Details
Gene Symbol: Iqub | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034130.1 | NP_001029302.1 | 8 - 9 | 1456 | 80 | |
GenBank mRNA | BC099086.1 | - | 8 - 9 | 1456 | 80 | |
DQ132434.1 | - | 8 - 9 | 1503 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
