Assay Details
Gene Symbol: Tac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001124768.1 | NP_001118240.1 | 1 - 2 | 90 | 77 | |
NM_001124769.1 | NP_001118241.1 | 1 - 2 | 90 | 77 | ||
NM_001124770.1 | NP_001118242.1 | 1 - 2 | 90 | 77 | ||
NM_012666.2 | NP_036798.1 | 1 - 2 | 90 | 77 | ||
GenBank mRNA | M34184.1 | - | 1 - 2 | 92 | 77 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the M34184.1, NM_001124768.1, NM_001124769.1, NM_001124770.1, NM_012666.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
