Assay Details
Gene Symbol: Trpv5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053787.2 | NP_446239.2 | 10 - 11 | 1421 | 114 | |
XM_006236376.2 | XP_006236438.1 | 8 - 9 | 997 | 114 | ||
GenBank mRNA | AB032019.1 | - | 10 - 11 | 1421 | 114 | |
AF209196.1 | - | 1372 | 114 | |||
AY762624.1 | - | 9 - 10 | 1109 | 114 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
