Assay Details
Gene Symbol: Napg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107384.1 | NP_001100854.1 | 5 - 6 | 484 | 96 | |
XM_006254829.2 | XP_006254891.1 | 6 - 7 | 476 | 96 | ||
GenBank mRNA | AY724541.1 | - | 6 - 7 | 381 | 96 | |
FQ211390.1 | - | 3 - 4 | 358 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
