Assay Details
Gene Symbol: Ccs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053425.1 | NP_445877.1 | 4 - 5 | 123 | ||
GenBank mRNA | AF255305.1 | - | 4 - 5 | 123 | ||
BC158586.1 | - | 4 - 5 | 437 | 123 | ||
EU340033.1 | - | 3 - 4 | 339 | 123 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
