Assay Details
Gene Symbol: Ccs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053425.1 | NP_445877.1 | 6 - 7 | 623 | 65 | |
GenBank mRNA | AF255305.1 | - | 6 - 7 | 623 | 65 | |
BC158586.1 | - | 6 - 7 | 583 | 65 | ||
EU340033.1 | - | 5 - 6 | 485 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
