Assay Details
Gene Symbol: LOC100911692 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006230595.2 | XP_006230657.1 | 8 - 9 | 2498 | 79 | |
XM_006230596.2 | XP_006230658.1 | 9 - 10 | 1862 | 79 | ||
XM_006230597.2 | XP_006230659.1 | 8 - 9 | 1750 | 79 |
Gene Symbol: Cracr2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127541.1 | NP_001121013.1 | 1125 | 79 | ||
XM_006230559.2 | XP_006230621.1 | 8 - 9 | 2498 | 79 | ||
XM_006230561.2 | XP_006230623.1 | 8 - 9 | 1750 | 79 | ||
GenBank mRNA | BC158864.1 | - | 1125 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
