Assay Details
Gene Symbol: Sigirr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024887.1 | NP_001020058.1 | 2 - 3 | 206 | 93 | |
XM_006230552.2 | XP_006230614.1 | 2 - 3 | 1040 | 93 | ||
XM_006230553.2 | XP_006230615.1 | 2 - 3 | 672 | 93 | ||
XM_008759988.1 | XP_008758210.1 | 1 - 2 | 2238 | 93 | ||
XM_008759989.1 | XP_008758211.1 | 2 - 3 | 731 | 93 | ||
XM_008759990.1 | XP_008758212.1 | 2 - 3 | 234 | 93 | ||
GenBank mRNA | BC097488.1 | - | 2 - 3 | 206 | 93 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC097488.1, NM_001024887.1, XM_006230552.2, XM_006230553.2, XM_008759988.1, XM_008759989.1, XM_008759990.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
