Assay Details
Gene Symbol: Sigirr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024887.1 | NP_001020058.1 | 3 - 4 | 410 | 83 | |
XM_006230552.2 | XP_006230614.1 | 3 - 4 | 1244 | 83 | ||
XM_006230553.2 | XP_006230615.1 | 3 - 4 | 876 | 83 | ||
XM_008759988.1 | XP_008758210.1 | 2 - 3 | 2442 | 83 | ||
XM_008759989.1 | XP_008758211.1 | 3 - 4 | 935 | 83 | ||
XM_008759990.1 | XP_008758212.1 | 3 - 4 | 438 | 83 | ||
GenBank mRNA | BC097488.1 | - | 3 - 4 | 410 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
