Assay Details
Gene Symbol: Sigirr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024887.1 | NP_001020058.1 | 5 - 6 | 676 | 76 | |
XM_006230552.2 | XP_006230614.1 | 5 - 6 | 1510 | 76 | ||
XM_006230553.2 | XP_006230615.1 | 5 - 6 | 1142 | 76 | ||
XM_008759988.1 | XP_008758210.1 | 4 - 5 | 2708 | 76 | ||
XM_008759989.1 | XP_008758211.1 | 5 - 6 | 1201 | 76 | ||
XM_008759990.1 | XP_008758212.1 | 5 - 6 | 704 | 76 | ||
GenBank mRNA | BC097488.1 | - | 5 - 6 | 676 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
