Assay Details
Gene Symbol: Klc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003749042.3 | XP_003749090.1 | 14 - 15 | 1971 | 108 | |
XM_003749043.3 | XP_003749091.1 | 14 - 15 | 1934 | 108 | ||
XM_006230934.1 | XP_006230996.1 | 14 - 15 | 2047 | 108 | ||
XM_006230935.2 | XP_006230997.1 | 14 - 15 | 1917 | 108 | ||
XM_006230936.1 | XP_006230998.1 | 14 - 15 | 1918 | 108 | ||
XM_006230937.2 | XP_006230999.1 | 14 - 15 | 1866 | 108 | ||
XM_008760196.1 | XP_008758418.1 | 13 - 14 | 2355 | 108 | ||
GenBank mRNA | BC166555.1 | - | 14 - 15 | 1939 | 108 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
