Assay Details
Gene Symbol: Klc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003749042.3 | XP_003749090.1 | 9 - 10 | 1459 | 76 | |
XM_003749043.3 | XP_003749091.1 | 9 - 10 | 1422 | 76 | ||
XM_006230934.1 | XP_006230996.1 | 9 - 10 | 1535 | 76 | ||
XM_006230935.2 | XP_006230997.1 | 9 - 10 | 1405 | 76 | ||
XM_006230936.1 | XP_006230998.1 | 9 - 10 | 1406 | 76 | ||
XM_006230937.2 | XP_006230999.1 | 9 - 10 | 1354 | 76 | ||
XM_008760196.1 | XP_008758418.1 | 8 - 9 | 1843 | 76 | ||
GenBank mRNA | BC166555.1 | - | 9 - 10 | 1427 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
