Assay Details
Gene Symbol: Dpf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108516.1 | NP_001101986.1 | 7 - 8 | 827 | 61 | |
XM_006230850.1 | XP_006230912.1 | 8 - 9 | 894 | 61 | ||
GenBank mRNA | BC166788.1 | - | 7 - 8 | 774 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
