Assay Details
Gene Symbol: Rin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139038.1 | NP_620607.1 | 1 - 2 | 291 | 123 | |
GenBank mRNA | U80076.1 | - | 1 - 2 | 291 | 123 |
Gene Symbol: LOC100911794 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003749037.3 | XP_003749085.1 | 1 - 2 | 573 | 123 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
