Assay Details
Gene Symbol: Rin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139038.1 | NP_620607.1 | 7 - 8 | 1795 | 68 | |
XM_008760073.1 | XP_008758295.1 | 2221 | 68 | |||
GenBank mRNA | U80076.1 | - | 7 - 8 | 1795 | 68 |
Gene Symbol: LOC100911794 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003749037.3 | XP_003749085.1 | 7 - 8 | 2077 | 68 | |
XM_008760195.1 | XP_008758417.1 | 6 - 7 | 2221 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
