Assay Details
Gene Symbol: Rce1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100585.1 | NP_001094055.1 | 6 - 7 | 702 | 76 | |
XM_008760136.1 | XP_008758358.1 | 6 - 7 | 642 | 76 | ||
GenBank mRNA | BC087629.1 | - | 6 - 7 | 689 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
