Assay Details
Gene Symbol: Map3k11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013150.1 | NP_001013168.1 | 6 - 7 | 2044 | 69 | |
GenBank mRNA | BC081952.1 | - | 6 - 7 | 2044 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
