Assay Details
Gene Symbol: Pdgfb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031524.1 | NP_113712.1 | 3 - 4 | 645 | 84 | |
GenBank mRNA | Z14117.1 | - | 3 - 4 | 226 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
