Assay Details
Gene Symbol: B4galnt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022860.2 | NP_074051.1 | 2 - 3 | 356 | 146 | |
XM_006241497.2 | XP_006241559.1 | 2 - 3 | 883 | 146 | ||
XM_008765419.1 | XP_008763641.1 | 2 - 3 | 2712 | 146 | ||
XM_008765420.1 | XP_008763642.1 | 1 - 2 | 1033 | 146 | ||
GenBank mRNA | BC081799.1 | - | 2 - 3 | 356 | 146 | |
D17809.1 | - | 2 - 3 | 245 | 146 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
