Assay Details
Gene Symbol: B4galnt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022860.2 | NP_074051.1 | 8 - 9 | 1143 | 93 | |
XM_006241497.2 | XP_006241559.1 | 8 - 9 | 1670 | 93 | ||
XM_008765419.1 | XP_008763641.1 | 8 - 9 | 3499 | 93 | ||
XM_008765420.1 | XP_008763642.1 | 7 - 8 | 1820 | 93 | ||
GenBank mRNA | BC081799.1 | - | 8 - 9 | 1143 | 93 | |
D17809.1 | - | 8 - 9 | 1032 | 93 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
