Assay Details
Gene Symbol: Stac3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130558.1 | NP_001124030.1 | 10 - 11 | 983 | 124 | |
XM_006241486.2 | XP_006241548.1 | 10 - 11 | 1496 | 124 | ||
XM_006241488.2 | XP_006241550.1 | 9 - 10 | 1428 | 124 | ||
XM_008765415.1 | XP_008763637.1 | 10 - 11 | 918 | 124 | ||
GenBank mRNA | FQ216646.1 | - | 9 - 10 | 807 | 124 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
