Assay Details
Gene Symbol: Srebf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033694.1 | NP_001028866.1 | 12 - 13 | 2499 | 58 | |
XM_006242079.2 | XP_006242141.1 | 12 - 13 | 2854 | 58 | ||
GenBank mRNA | BC101902.1 | - | 12 - 13 | 2499 | 58 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
