Assay Details
Gene Symbol: Srebf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033694.1 | NP_001028866.1 | 18 - 19 | 3321 | 149 | |
XM_006242079.2 | XP_006242141.1 | 18 - 19 | 3676 | 149 | ||
GenBank mRNA | BC101902.1 | - | 18 - 19 | 3321 | 149 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription by RNA polymerase II
transcription, DNA-templated
regulation of transcription, DNA-templated
spermatogenesis
aging
cholesterol metabolic process
lipid biosynthetic process
response to xenobiotic stimulus
response to hormone
response to lead ion
positive regulation of cholesterol storage
response to lipid
cholesterol homeostasis
positive regulation of transcription, DNA-templated
positive regulation of transcription by RNA polymerase II
cellular response to low-density lipoprotein particle stimulus
negative regulation of cholesterol efflux
C-8 sterol isomerase activity
transcription cis-regulatory region binding
RNA polymerase II cis-regulatory region sequence-specific DNA binding
DNA binding
chromatin binding
DNA-binding transcription factor activity
protein C-terminus binding
sequence-specific DNA binding
protein dimerization activity
E-box binding
negative regulation of transcription by RNA polymerase II
transcription, DNA-templated
regulation of transcription, DNA-templated
spermatogenesis
aging
cholesterol metabolic process
lipid biosynthetic process
response to xenobiotic stimulus
response to hormone
response to lead ion
positive regulation of cholesterol storage
response to lipid
cholesterol homeostasis
positive regulation of transcription, DNA-templated
positive regulation of transcription by RNA polymerase II
cellular response to low-density lipoprotein particle stimulus
negative regulation of cholesterol efflux
C-8 sterol isomerase activity
transcription cis-regulatory region binding
RNA polymerase II cis-regulatory region sequence-specific DNA binding
DNA binding
chromatin binding
DNA-binding transcription factor activity
protein C-terminus binding
sequence-specific DNA binding
protein dimerization activity
E-box binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P092209 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 167441 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 274735 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 412321 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 914009 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 233779 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 85083 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 354574 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 757554 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 801576 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 850146 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 491706 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 356929 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 17127 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 117889 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 147691 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 425368 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 378729 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 655960 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 736116 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 220075 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 630130 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 581356 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 41305 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 120593 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 764612 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 691071 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 467454 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 707239 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 440633 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 163346 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 336121 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 740889 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 219593 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 271975 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 185101 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 629164 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 601339 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 877813 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 140528 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 114413 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 73574 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 166948 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 48932 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 138063 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 162855 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 173180 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 141639 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 126503 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 108625 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P092209 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P092209 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P092209 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P382747 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P504215 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 61677 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 12857 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 28946 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 98687 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 49777 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 100726 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 72855 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10898092 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10898095 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10898100 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10898097 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10898105 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10898113 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10898112 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10898111 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10898096 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10898104 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10898114 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10898108 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10898107 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10898116 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10898102 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10898101 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10898099 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10898098 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10898110 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10898109 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10898115 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10898094 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10898106 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10898093 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10898091 | - | - |
Rat Genome 230 2.0 Array | 1371979_PM_at | - | - |
Rat Genome 230 2.0 Array | 1377489_PM_at | - | - |
Rat Genome 230 2.0 Array | 1377489_at | - | - |
Rat Genome 230 2.0 Array | 1371979_at | - | - |
Affymetrix HT Rat Focus Array | 1371979_at | - | - |
Affymetrix HT Rat Focus Array | 1371979_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1377489_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1377489_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0700011652.rn.1 | TC0700001520.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0700011653.rn.1 | TC0700001520.rn.1 | RTA TC |
Affymetrix Rat Gene 1.0 ST Array | 10898091 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22160023 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1354176 | - | - |
