Assay Details
Gene Symbol: Cpsf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006241372.1 | XP_006241434.1 | 10 - 11 | 1847 | 91 | |
XM_006241374.1 | XP_006241436.1 | 10 - 11 | 1844 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
