Assay Details
Gene Symbol: Mdm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108099.1 | NP_001101569.1 | 1 - 2 | 169 | 131 | |
XM_006241382.1 | XP_006241444.1 | 1 - 2 | 459 | 131 | ||
XM_008765396.1 | XP_008763618.1 | 1 - 2 | 459 | 131 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_001108099.1, XM_006241382.1, XM_008765396.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
