Assay Details
Gene Symbol: Mdm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108099.1 | NP_001101569.1 | 10 - 11 | 1045 | 134 | |
XM_006241382.1 | XP_006241444.1 | 11 - 12 | 1410 | 134 | ||
XM_006241383.1 | XP_006241445.1 | 11 - 12 | 974 | 134 | ||
XM_008765396.1 | XP_008763618.1 | 12 - 13 | 1482 | 134 | ||
XM_008765397.1 | XP_008763619.1 | 12 - 13 | 1046 | 134 | ||
XM_008765399.1 | XP_008763621.1 | 8 - 9 | 792 | 134 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
