Assay Details
Gene Symbol: Dram1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173427.1 | NP_001166898.1 | 6 - 7 | 784 | 122 | |
GenBank mRNA | BC168648.1 | - | 6 - 7 | 784 | 122 | |
FQ211820.1 | - | 6 - 7 | 912 | 122 | ||
FQ223303.1 | - | 6 - 7 | 844 | 122 | ||
FQ229264.1 | - | 6 - 7 | 845 | 122 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
