Assay Details
Gene Symbol: Kitlg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021843.4 | NP_068615.1 | 1 - 2 | 186 | 108 | |
NM_021844.2 | NP_068616.1 | 1 - 2 | 186 | 108 | ||
XM_008765339.1 | XP_008763561.1 | 1 - 2 | 230 | 108 | ||
XM_008765340.1 | XP_008763562.1 | 1 - 2 | 230 | 108 | ||
GenBank mRNA | AB105879.1 | - | 1 - 2 | 186 | 108 | |
CB751462.1 | - | 1 - 2 | 186 | 108 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AB105879.1, NM_021843.4, NM_021844.2, XM_008765339.1, XM_008765340.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
