Assay Details
Gene Symbol: Kitlg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021843.4 | NP_068615.1 | 8 - 9 | 949 | 136 | |
NM_021844.2 | NP_068616.1 | 7 - 8 | 865 | 136 | ||
GenBank mRNA | AB105879.1 | - | 7 - 8 | 865 | 136 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
