Assay Details
Gene Symbol: Kitlg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021843.4 | NP_068615.1 | 9 - 10 | 1039 | 140 | |
NM_021844.2 | NP_068616.1 | 8 - 9 | 955 | 140 | ||
GenBank mRNA | AB105879.1 | - | 8 - 9 | 955 | 140 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
