Assay Details
Gene Symbol: Nudt4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053598.1 | NP_446050.1 | 4 - 5 | 447 | 113 | |
XM_006241282.2 | XP_006241344.1 | 4 - 5 | 641 | 113 | ||
GenBank mRNA | AF253473.1 | - | 4 - 5 | 447 | 113 | |
FQ223117.1 | - | 4 - 5 | 532 | 113 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
