Assay Details
Gene Symbol: Nap1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053561.2 | NP_446013.2 | 13 - 14 | 1250 | 111 | |
XM_006241342.1 | XP_006241404.1 | 13 - 14 | 1264 | 111 | ||
XM_006241344.2 | XP_006241406.1 | 11 - 12 | 1001 | 111 | ||
GenBank mRNA | AF062594.1 | - | 13 - 14 | 1232 | 111 | |
CA511505.1 | - | 2 - 3 | 81 | 111 | ||
CF110129.1 | - | 8 - 9 | 685 | 111 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
