Assay Details
Gene Symbol: Cpm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108098.1 | NP_001101568.1 | 8 - 9 | 1121 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P669544 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 27993 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 556034 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 177441 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 204108 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 184200 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 528403 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 782539 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 214599 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 341195 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 884258 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 383304 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 384789 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 802129 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 868597 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 211960 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 512071 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 537001 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 94561 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 324007 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 201182 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 295898 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 964159 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 6516 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 911961 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 168060 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 802180 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 163962 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 226890 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 849822 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 633121 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 458971 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 188324 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 344862 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 868925 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 607188 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 170787 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 897050 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 560874 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 427407 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 525 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 868131 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 889715 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 131249 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 16063 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 846230 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 215940 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 24166 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 46548 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 68775 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 133681 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 42711 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 142756 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P669544 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P669544 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P669544 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P669544 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 83586 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10895596 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10895595 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10895597 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10895590 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10895592 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10895594 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10895593 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10895591 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10895589 | - | - |
Rat Genome 230 2.0 Array | 1394548_at | - | - |
Rat Genome 230 2.0 Array | 1394548_PM_at | - | - |
Rat Genome 230 2.0 Array | 1391656_PM_at | - | - |
Rat Genome 230 2.0 Array | 1391656_at | - | - |
Affymetrix HT Rat Focus Array | 1391656_at | - | - |
Affymetrix HT Rat Focus Array | 1391656_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1394548_at | - | - |
Affymetrix HT Rat Focus Array | 1394548_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0700005873.rn.1 | TC0700000761.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0700005874.rn.1 | TC0700000761.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0700005872.rn.1 | TC0700000761.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0700005876.rn.1 | TC0700000761.rn.1 | RTA TC |
Rat Expression Set 230 | 1391656_at | - | - |
Rat Expression Set 230 | 1391656_PM_at | - | - |
Rat Expression Set 230 | 1394548_at | - | - |
Rat Expression Set 230 | 1394548_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10895589 | - | - |
Rat Genome U34 Set | rc_AI009946_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22270903 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1373155 | - | - |
